Learn More
Medchemexpress LLC HY-N0356 10mg Medchemexpress, (-)-Catechin gallate CAS:130405-40-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN035610MG
Encompass_Preferred
Medchemexpress, HY-N0356 10mg (-)-Catechin gallate CAS:130405-40-2 (-)-Catechin gallate is a minor constituent in green tea catechins. (-)-Catechin gallate inhibits the activity of COX-1 and COX-2 enzymes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3535