Learn More
Medchemexpress LLC (±)-Catechin (rel-Cianidanol; rel-Catechuic acid) | 7295-85-4 | 99.5% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB1890500MG
Encompass_Preferred
(±)-Catechin (rel-Cianidanol) is the racemate of the green tea polyphenol Catechin. Catechin has anticancer activity and induces apoptosis. (±)-Catechin has two forms, (+)-Catechin and its enantiomer (-)-Catechin. (+)-Catechin inhibits cyclooxygenase-1 (COX-1) with an IC50 of 1.4 μM. (-)-Catechin can effectively promote hBM-MSC adipocyte differentiation and increase adiponectin and PPARγ levels. (±)-Catechin has anti-tumor, anti-obesity, anti-diabetic, anti-cardiovascular, anti-infectious, hepatoprotective and neuroprotective effects.
  • Induces adipocyte differentiation of human bone marrow mesenchymal stem cells (hBM-MSC)
  • Increases adiponectin levels
  • Increases PPARγ transcription
Catalog No. 50-004-12398