Learn More
Cayman Chemical -CatechInhydrate 1g
SDP
Supplier:  Cayman Chemical 291751 G
Encompass_Preferred
A polyphenol flavonoid; reduces iron- and ascorbate-induced lipid peroxidation of isolated bovine brain phospholipid liposomes (IC50 = 20 µM); inhibits HCV RdRp by 17.7% in an enzyme assay
Catalog No. 50-294-0960