Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-catenin-IN-2 | 1458664-10-2 | 99.6% | 255.29 | 50 MG
SDP

Supplier:  Medchemexpress LLC HY13646450MG

Encompass_Preferred

β-catenin-IN-2 is a potent β-catenin inhibitor, also known as Compound H1B1. It is utilized in the study of colorectal cancer, where it suppresses the transcriptional activity of β-catenin/Tcf-4 in HCT116 cells in a dose-dependent manner at concentrations of 10-40 μM.

  • Solid appearance with pink to red color
  • High purity of 99.6%
  • Molecular weight of 255.29
  • Formula: C15H14FN3
  • Inhibits β-catenin/Tcf-4 transcriptional activity
  • Used in colorectal cancer research

Catalog No. 50-003-52328


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.