Learn More
ATCC CATH.a Neuronal Cell Line (mouse brain) - ATCC CRL-3595
Supplier:  ATCC CRL3595
Encompass
CATH.a is a transformed mouse neuronal cell line derived from the brain of a transgenic mouse. It is used in neuroscience and toxicology research to investigate neuronal properties and neurotoxic responses.
  • Derived from mouse brain tissue.
  • Neuron morphology indicative of neuronal phenotype.
  • Transformed cell line suitable for in vitro studies.
  • Commonly used in neuroscience and toxicology research.
  • Useful for studies of catecholaminergic neuronal properties.
Catalog No. NC3835360