Learn More
Med Vet International Introcan Safety Fep IV Catheter, 20g X 1.25in., Straight, 50/Box
Supplier:  Med Vet International BMG4252535Z
Encompass_Preferred
The Introcan Safety 20g x 1.25in. IV Catheter offers a passive safety feature that is activated automatically and cannot be bypassed. The easy-to-use safety IV Catheter incorporates a separate needle and catheter flashback that gives you the added confidence of a successful placement every time. The sharp needle features a universal bevel geometry that allows a wider choice of insertion angles, making the catheter easy to adapt to and vein puncture more comfortable for the patient. The 20g x 1.25in. Safety IV Catheter is not made with DEHP, Natural Rubber, or with PVC. This safety IV Catheter is Sterile.
Catalog No. 50-283-0443