Learn More
Medchemexpress LLC Cathepsin C/DPPI protein, mouse (inactive, His-Myc) | >95.0% | 10UG
SDP
Supplier:  Medchemexpress LLC HYP70124710UG
Encompass_Preferred
Recombinant inactive mouse Cathepsin C/DPPI protein with N-terminal His and C-terminal Myc tags, supplied as a lyophilized powder. The protein exhibits both exopeptidase and endopeptidase activities and can activate serine proteases such as elastase, cathepsin G, and granzymes A and B. It is expressed in E. coli and purified to high purity for research applications.
Catalog No. 50-001-76417