Learn More
Medchemexpress LLC Cathepsin C/DPPI protein, mouse (inactive, His-Myc) | >95.0% | 500UG
SDP
Supplier:  Medchemexpress LLC HYP701247500UG
Encompass_Preferred
Recombinant mouse Cathepsin C/DPPI protein (inactive, His-Myc) provided lyophilized for research applications. The preparation is expressed in E. coli, formulated in 10 mM Tris-HCl, 1 mM EDTA, 6% trehalose (pH 8.0), and exhibits >95.0% purity by reducing SDS-PAGE. Supplied in multiple pack sizes and intended for storage frozen to preserve integrity.
  • Inactive protein suitable for detection and structural studies.
  • N-terminal 10xHis and C-terminal Myc tags for purification and detection.
  • High purity (>95.0%) confirmed by reducing SDS-PAGE.
  • Lyophilized formulation with trehalose for enhanced stability.
  • Expressed in E. coli for cost-effective production.
  • Stable when stored at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.
Catalog No. 50-001-75837