Learn More
Medchemexpress LLC Cathepsin D (CTSD) | 9025-26-7 | 10 U
SDP
Supplier:  Medchemexpress LLC HYP275010U
Encompass_Preferred
Cathepsin D (CTSD) is a lysosomal aspartic protease that regulates lysosomal protease activity. Its loss can lead to lysosomal dysfunction and the accumulation of cellular proteins associated with neurodegenerative diseases. This product is also involved in breast cancer metastasis and is used in research on diseases like breast cancer and stroke.
  • Lysosomal aspartic protease
  • Regulates lysosomal protease activity
  • Involved in breast cancer metastasis
  • Used in research on neurodegenerative diseases
  • Used in research on breast cancer and stroke
Catalog No. 50-004-00378