Learn More
Medchemexpress LLC Cathepsin G inhibitor I | 429676-93-7 | MFCD28137715 | >99.0% | 620.63 g/mol | C36H33N2O6P | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1033515MG
Encompass_Preferred
Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G used in biochemical research to modulate protease activity. It is supplied as a solid or as a DMSO solution and is intended for laboratory research use.
  • Selective, reversible, competitive inhibition of cathepsin G.
  • Potent activity: IC50 53 nM, Ki 63 nM.
  • High purity (>99.0%).
  • Available in small pack sizes and as a 10 mM DMSO solution.
  • Suitable for biochemical and cellular studies to probe cathepsin G function.
Catalog No. 50-002-45057