Learn More
Medchemexpress LLC Cathepsin G inhibitor I | 429676-93-7 | MFCD28137715 | ≥99.0% | 620.63 g·mol⁻¹ | C36H33N2O6P | 50 MG

Supplier: Medchemexpress LLC HY10335150MG
Cathepsin G Inhibitor I is a selective, reversible, competitive, non-peptide inhibitor of cathepsin G used in biochemical and cellular research to probe cathepsin G-mediated processes. It is supplied as a solid and as a DMSO solution and is formulated for assay-ready use.
- Selective, reversible, competitive inhibitor of cathepsin G (IC50 = 53 nM; Ki = 63 nM).
- High purity suitable for research use (≥99.0%).
- Soluble in DMSO at 50 mg/mL with ultrasonic warming for in vitro applications.
- Available as a solid and as a 10 mM solution in DMSO for convenience in assays.
- Storage guidance provided for solid and solution forms to preserve stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.