Learn More
Apexbio Technology LLC Cathepsin G Inhibitor I(Synonyms: Cathepsin G inhibitor, Cathepsin G inhibitor I, CatG inhibitor, CatG inhibitor I), 5mg, CAS: 429676-93-7.
SDP
Supplier:  Apexbio Technology LLC A81745
Encompass_Preferred
Cathepsin G Inhibitor I (CAS 429676-93-7) is a selective reversible and competitive nonpeptide inhibitor targeting the serine protease Cathepsin G (EC 3 4 21 20) an enzyme encoded by the human CTSG gene that functions in proteolytic cleavage processes In vitro this compound exhibits potent inhibitory activity against Cathepsin G with reported IC50 and Ki values of 53 12 nM and 63 14 nM respectively while displaying weaker inhibition against chymotrypsin (Ki 1 5 0 2 M) and minimal inhibitory effect ( 50% inhibition at 100 M) on several other serine proteases Cathepsin G Inhibitor I is currently utilized for preclinical research and enzymatic studies
Catalog No. 50-000-61035