Learn More
Cayman Chemical CathepsIn G InhIbItr I 500ug
SDP
Supplier:  Cayman Chemical 14928500 UG
Encompass_Preferred
A cathepsin G inhibitor (IC50 = 53 nM); selective for cathepsin G over thrombin, Factor Xa, Factor IXa, plasmin, trypsin, and PR3 (IC50s = >100 µM for all), but also weakly inhibits chymotrypsin; inhibits cathepsin G-induced activation of IL-36 in neutrophil degranulates at 10 µM; reduces CD4+ T cell-induced secretion of IFN-γ and IL-17 in B cells incubated with TTC and also reduces TTC presentation in B cells at 10 µM; decreases the size of neuromyelitis optica lesions and reduces the number of perivascular neutrophils when used in combination with sivelestat in a mouse model of IgG-induced brain injury at 5 UG/animal
Catalog No. 50-297-0940