Learn More
Medchemexpress LLC Cathepsin Inhibitor 2 | 1017931-53-1 | 421.38 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYU003775MG
Encompass_Preferred
Cathepsin Inhibitor 2 is a potent Cathepsin S inhibitor, demonstrating a Ki of <20 nM. It also potently inhibits Cathepsin F, Cathepsin L, and Cathepsin B with Kis of 21-500 nM. This product is for research use only.
  • Potent Cathepsin S inhibitor (Ki: <20 nM)
  • Inhibits Cathepsin F, Cathepsin L, and Cathepsin B (Kis: 21-500 nM)
  • Suitable for research use only
Catalog No. 50-004-49632