Learn More
Medchemexpress LLC Cathepsin K inhibitor 6 | 325775-42-6 | 98.2% | 251.3 g/mol | C11H13N3O2S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY14777410MG
Encompass_Preferred
Cathepsin K inhibitor 6 is a small-molecule research compound that potently inhibits cathepsin K (IC50 17 nM) and shows activity against related cathepsin isoforms. The compound is supplied as a solid for laboratory use; molecular formula C11H13N3O2S, molecular weight 251.3 g/mol, CAS 325775-42-6.
Catalog No. 50-002-86975