Learn More
Sino Biological Recombinant Anti-Cathepsin L Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10486R221100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Cathepsin L / CTSL / CTSL1 (rh Cathepsin L / CTSL / CTSL1; Catalog#10486-H08H; NP_001903.1; Met1-Val333).
Catalog No. 50-224-8087