Learn More
Medchemexpress LLC CATPB 1mg | 1322598-09-3 | 1 MG

Supplier: Medchemexpress LLC HY1162631MG
CATPB is a potent, selective antagonist of free fatty acid receptor 2 (FFA2/GPR43) intended for research use. It is supplied as a white to off-white solid with high purity and defined storage conditions, provided in a 1 mg quantity.
- Potent, selective antagonist of FFA2 (GPR43)
- Molecular formula C19H17ClF3NO3; molecular weight 399.79
- Purity 99.7%
- Appearance: white to off-white solid
- Storage: powder -20°C for long term; 4°C for short term
- Intended for research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.