Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cauloside A | 17184-21-3 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN35571MG

Encompass_Preferred

Cauloside A (Leontoside A) is a saponin isolated from Dipsacus asper roots. It has potent antifungal activity.

  • Isolated from Dipsacus asper roots
  • Potent antifungal activity
  • Purity: 99.55%
  • Molecular weight: 604.81
  • Chemical formula: C35H56O8
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, triterpenes
  • Solubility in DMSO: 100 mg/mL (165.34 mM)

Catalog No. 50-004-19397


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.