Learn More
Medchemexpress LLC Cauloside A | 17184-21-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN35575MG
Encompass_Preferred
Cauloside A (Leontoside A) is a saponin isolated from Dipsacus asper roots and possesses potent antifungal activity.
  • Saponin isolated from Dipsacus asper roots.
  • Exhibits potent antifungal activity.
  • Exhibits cytotoxicity against human A549, DLD-1, HL-60, HeLa, HepG2, and U-87MG cells.
  • Shows anti-inflammatory activity in mouse RAW264.7 cells.
Catalog No. 50-004-19436