Learn More
Fine Science Tools Small Vessel Cauterizer Kit, Stainless Steel, 13.5 cm, 100 g
Supplier: Fine Science Tools 1800000
Product: Small Vessel Cauterizer Kit/ Alloy : Stainless Steel/ Handle Weight : 100g/ Handle Length : 13.5cm/ Autoclave Safe: Yes/ Unit : Each