Learn More
Medchemexpress LLC HY-119373 10mg Medchemexpress, Cav 2.2 blocker 1 CAS:1567335-29-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11937310MG
Encompass_Preferred
Medchemexpress, HY-119373 10mg Cav 2.2 blocker 1 CAS:1567335-29-8 Cav 2.2 blocker 1 (compound 9) is a N-type calcium channel (Cav 2.2) blocker for the treatment of pain, with an IC50 of 1 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9531