Learn More
Medchemexpress LLC Cavosonstat | 1371587-51-7 | 99.4% | 299.71 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10902750MG
Encompass_Preferred
Cavosonstat (N91115) is an orally active S-nitrosoglutathione reductase (GSNOR) inhibitor. It is also a CFTR stabilizer, and can be used for cystic fibrosis research. In vitro, Cavosonstat, a CFTR modulator, promotes cystic fibrosis transmembrane conductance regulator (CFTR) maturation and plasma membrane stability, with a mechanism of action complementary to correctors and potentiators.
  • Orally active S-nitrosoglutathione reductase (GSNOR) inhibitor.
  • CFTR stabilizer for cystic fibrosis research.
  • Promotes cystic fibrosis transmembrane conductance regulator (CFTR) maturation and plasma membrane stability.
  • Mechanism of action complementary to CFTR correctors and potentiators.
Catalog No. 50-003-44652