Learn More
Medchemexpress LLC Cavosonstat | 1371587-51-7 | 99.4% | 299.71 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10902710MG
Encompass_Preferred
Cavosonstat is an orally active S-nitrosoglutathione reductase (GSNOR) inhibitor. It is also a CFTR stabilizer and can be used for cystic fibrosis research. As a CFTR modulator, it promotes cystic fibrosis transmembrane conductance regulator (CFTR) maturation and plasma membrane stability, with a mechanism of action complementary to CFTR correctors and potentiators.
  • Orally active S-nitrosoglutathione reductase (GSNOR) inhibitor.
  • CFTR stabilizer.
  • Can be used for cystic fibrosis research.
  • Promotes CFTR maturation and plasma membrane stability.
  • Mechanism of action complementary to CFTR correctors and potentiators.
  • Appearance: Solid.
  • Color: Light yellow to yellow.
Catalog No. 50-003-45419