Learn More
Medchemexpress LLC Cavosonstat | 1371587-51-7 | 99.4% | 299.71 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY109027
Encompass_Preferred
Cavosonstat (N91115) is an orally active S-nitrosoglutathione reductase (GSNOR) inhibitor. It also acts as a CFTR stabilizer and can be used in research related to cystic fibrosis. As a CFTR modulator, cavosonstat promotes CFTR maturation and plasma membrane stability, complementing the action of CFTR correctors and potentiators.
  • Orally active S-nitrosoglutathione reductase (GSNOR) inhibitor.
  • CFTR stabilizer.
  • Promotes CFTR maturation and plasma membrane stability.
  • Mechanism of action is complementary to CFTR correctors and potentiators.
Catalog No. 50-003-47021