Learn More
Medchemexpress LLC Cavosonstat | 1371587-51-7 | 99.4% | 299.71 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY109027100MG
Encompass_Preferred
Cavosonstat (N91115) is an orally active S-nitrosoglutathione reductase (GSNOR) inhibitor. It also acts as a CFTR stabilizer and is used for cystic fibrosis research. As a CFTR modulator, Cavosonstat promotes the maturation and plasma membrane stability of the cystic fibrosis transmembrane conductance regulator (CFTR), offering a mechanism of action that complements CFTR correctors and potentiators.
  • Orally active S-nitrosoglutathione reductase (GSNOR) inhibitor
  • CFTR stabilizer
  • Used for cystic fibrosis research
  • Promotes CFTR maturation and plasma membrane stability
  • Complements CFTR correctors and potentiators
Catalog No. 50-003-46331