Learn More
Cayman Chemical CAY10397, 78028-01-0, 5 mg
SDP
Supplier:  Cayman Chemical 701305
Encompass_Preferred
Molecular Formula C17H16N2O5, Purity ≥98%, Formula Weight 328.3, Synonyms: 5-[[4-(ethoxycarbonyl)phenyl]azo]-2-hydroxy-benzeneacetic acid, CK47A; A selective inhibitor of 15-hydroxy PGDH (IC50 = ~10 M).
Catalog No. NC1573094