Learn More
Medchemexpress LLC HY-121537 5mg Medchemexpress, CAY10404 CAS:340267-36-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1215375MG
Encompass_Preferred
Medchemexpress, HY-121537 5mg CAY10404 CAS:340267-36-9 CAY10404 is a potent and selective cyclooxygenase-2 (COX-2) inhibitor with an IC50 of 1 nM and a selectivity index (SI; COX-1 IC50/COX-2 IC50) of >500000. CAY10404 is a potent PKB/Akt and MAPK signaling pathways inhibitor and induces apoptosis in non-small cell lung cancer (NSCLC) cells. CAY10404, a diarylisoxazole, has good analgesic, anti-inflammatory, and anti-cancer activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0972