Learn More
TARGETMOL CHEMICALS INC CAY10434 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T378255MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. CAY10434 is an active inhibitor of CYP4A hydroxylase with an IC50 value of 8.8 nM in human kidney microsomal test. The maximum contractile value (Emax) of CAY 10434 was 6764 mg in improving the contractile process of angiotensin II. purity: 99%
Catalog No. 50-258-0121