Learn More
Cayman Chemical CAY10434 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 100050675 MG
Encompass_Preferred
A selective inhibitor of the 20-HETE synthase CYP4A11 (IC50 = 8.8 nM human renal microsomes)
Catalog No. 50-289-1142