Learn More
Cayman Chemical CAY10486 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 1000645250 MG
Encompass_Preferred
An inhibitor of ACAT; inhibits human ACAT-1 and ACAT-2 (IC50s = ~60 µM for both); inhibits copper-mediated oxidation of LDL by about 20% at 3 µM
Catalog No. 50-288-8047