Learn More
Abcam CAY10498, A3 adenosine receptor antagonist, 1MG
Supplier:  Abcam AB1444551MG
Encompass_Preferred
MW 320.4 Da, Purity >98%. Potent, selective A3 adenosine receptor antagonist (Ki = 37 nM). Reversine (ab120921) analog.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5700