Learn More
Cayman Chemical CAY10565 Neurochemicals
SDP
Supplier:  Cayman Chemical 100105285 MG
Encompass_Preferred
A S-nitrosothiol that acts as an NO donor under acidic conditions; decomposes with a half-life of 130 minutes in 0.1 M phosphate buffer, pH 5.0, at 37°C; relaxes phenylephrine-constricted rat aortic strips 71% and 44% at pH 6.0 and 7.4, respectively
Catalog No. 50-288-7225