Learn More
Selleck Chemical LLC CAY10566-E1227-1G
SDP
Supplier:  Selleck Chemical LLC E12271G
Encompass_Preferred
CAY10566 is a potent orally bioavailable and selective inhibitor of stearoyl-CoA desaturase1 (SCD1) with IC50 of 4 5 and 26 nM in mouse and human enzymatic assays respectively It also demonstrates excellent cellular activity in blocking the conversion of saturated long-chain fatty acid-CoAs (LCFA-CoAs) to monounsaturated LCFA-CoAs in HepG2 cells
Catalog No. 50-313-7990