Learn More
Cayman Chemical CAY10567 Inhibitors
SDP
Supplier:  Cayman Chemical 10010233500 MG
Encompass_Preferred
An inhibitor of Akt1 translocation that also inhibits hepatitis C virus (HCV) NS5B RNA-dependent RNA polymerase (RdRp) (IC50 = 79 µM)
Catalog No. 50-288-8661