Learn More
Cayman Chemical CAY10573 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 10008846500 UG
Encompass_Preferred
A PPAR agonist that displays potent binding at PPARα, γ, and δ (IC50s = 113, 50, 223 nM, respectively) and potently transactivates PPARα, γ, and δ (EC50s = 8, 70, 500 nM, respectively); demonstrates stronger binding and functional activity for PPARγ than rosiglitazone
Catalog No. 50-289-1206