Learn More
Abcam CAY10585, HIF-1alpha inhibitor, 25MG
Supplier:  Abcam AB14442225MG
Encompass_Preferred
MW 435.5 Da, Purity >97%. HIF-1α inhibitor (IC₅₀ values are 2.6 and 0.7 μM for Hep3B and AGS cell lines respectively). Suppresses transcription of HIF-1 target genes VEGF and erythropoietin at 10 μM. Attenuates hypoxia-induced ANP secretion. Blunts BMP9-induced osteogenic signaling in MSCs.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5760