Learn More
Cayman Chemical CAY10591 Fatty Acids and Deriv
SDP
Supplier:  Cayman Chemical 100097971 MG
Encompass_Preferred
An activator of SIRT1 that decreases TNF-α levels from 325 pg/ml (control) to 104 and 53 pg/ml at 20 and 60 µM, respectively; exhibits a significant dose-dependent effect on fat mobilization in differentiated adipocytes
Catalog No. 50-289-1024