Learn More
TARGETMOL CHEMICALS INC CAY10602 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T406250MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CAY10602 is a SIRT1 activator. CAY10602 was derived from high throughput screening for compounds that increase SIRT1-mediated deacetylation of a SIRT1-specific substrate. Functional assays show that CAY10602 dose-dependently suppresses the NF-(kappa)B-dependent induction of TNF-(alpha) by lipopolysaccharide in THP-1 cells with approximately 75% inhibition achieved at 60 uM without cytotoxicity. purity: 98%
Catalog No. 50-257-9979