Learn More
Medchemexpress LLC HY-18613 1mg , CAY10603 CAS:1045792-66-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY186131MG
Encompass_Preferred
HY-18613 1mg CAY10603 CAS: 1045792-66-2 CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM; CAY10603 (BML-281) also inhibits HDAC1, HDAC2, HDAC3, HDAC8, HDAC10, with IC50s of 271, 252, 0.42, 6851, 90.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0217