Learn More
Selleck Chemical LLC CAY10603 5mg 1045792-66-2
SDP
Supplier:  Selleck Chemical LLC S75965MG
Encompass_Preferred
CAY10603 is a potent and selective HDAC6 inhibitor with IC50 of 2 pM, >200-fold selectivity over other HDACs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5003