Learn More
Cayman Chemical CAY10740hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 283205 MG
Encompass_Preferred
An analog of GSK484; decreases the viability of 4T1 cells by 85 and 98% at 5 and 10 μM, respectively, which is more than five-fold as effective as GSK484
Catalog No. 50-294-0394