Learn More
Medchemexpress LLC HY-101979A 10mg Medchemexpress, CB-1158 (dihydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101979A10MG
Encompass_Preferred
Medchemexpress, HY-101979A 10mg CB-1158 (dihydrochloride) CAS:0 CB-1158 dihydrochloride (INCB01158 dihydrochloride) is a potent and orally active inhibitor of arginase, with IC50s of 86 nM and 296 nM for recombinant human arginase 1 and recombinant human arginase 2, respectively. Immuno-oncology agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8694