Learn More
Abcam CB-1158 (Numidargistat), 1MG
Supplier:  Abcam AB2853881MG
Encompass_Preferred
MW 287.12 Da, Purity >=97%. Potent, selective inhibitor of human arginase I (IC₅₀ = 98 nM), an immune-mediated anti-tumor agent. Displays potential for anti-tumor activity in renal cell cancer, breast cancer, non-small cell lung cancer, acute myeloid leukemia, etc.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5512