Learn More
Medchemexpress LLC HY-12861 10mg Medchemexpress, CB-5083 CAS:1542705-92-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1286110MG
Encompass_Preferred
Medchemexpress, HY-12861 10mg CB-5083 CAS:1542705-92-9 CB-5083 is a potent, selective and orally bioavailable p97 inhibitor with an IC50 value of 11 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8922