Learn More
TARGETMOL CHEMICALS INC CB-5083 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T679610MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CB-5083 is a potent selective and orally bioavailable p97 AAA ATPase inhibitor ( IC50=11 nM). purity: 99%
Catalog No. 50-257-6452