Learn More
Selleck Chemical LLC CB-5083
SDP
Supplier:  Selleck Chemical LLC S8101100MG
Encompass_Preferred
CB-5083 is a potent selective and orally bioavailable p97 AAA ATPase inhibitor with IC50 of 11 nM Phase 1
Catalog No. 50-000-00108