Learn More
Apexbio Technology LLC CB-5083 1542705-92-9 50mg
SDP
Supplier:  Apexbio Technology LLC B603250
Encompass_Preferred
CB-5083 (CAS 1542705-92-9) is an orally bioavailable small-molecule inhibitor targeting the AAA ATPase p97 (also termed valosin-containing protein VCP) P97 functions in diverse cellular processes related to protein homeostasis including membrane fusion intracellular trafficking and degradation pathways CB-5083 selectively inhibits the second ATPase domain of p97 (IC50 15 4 nM) promoting cytoplasmic protein degradation and triggering apoptosis in cultured cancer cell lines such as A549 and HCT116 In vivo orally administered CB-5083 demonstrates significant reduction of tumor growth in xenograft models The compound is being investigated clinically for treatment of solid tumors and multiple myeloma
Catalog No. 50-000-62689