Learn More
TARGETMOL CHEMICALS INC CB-6644 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T106935MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CB-6644 is a selective inhibitor of the RUVBL1/2 complex and blocks the ATPase activity of RUVBL1/2 (IC50 = 15 nM). purity: 98%
Catalog No. 50-258-1689