Learn More
Cayman Chemical CB13 Other Receptor Modulators
SDP
Supplier:  Cayman Chemical 1001039810 MG
Encompass_Preferred
A dual agonist of the CB1 (IC50 = 15 nM) and CB2 (IC50 = 98 nM) receptors; potently blocks CB1-dependent neuropathic mechanical hyperalgesia in rats, shows limited brain penetration, and exhibits good oral bioavailability
Catalog No. 50-288-9223