Learn More
Cayman Chemical CB25 Neurochemicals
SDP
Supplier:  Cayman Chemical 100101161 MG
Encompass_Preferred
A stable analog of Δ9-THC; exhibits high affinity for the CB1 and CB2 receptors with Ki values of 5.2 and 13 nM, respectively, also it behaves as an inverse agonist for the CB1 receptor as assessed in a cyclic AMP functional assay
Catalog No. 50-289-0322